Product Details
- SNP ID
-
rs2014459
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52276496 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGGATGTGAGGCAGAGGGTCCC[C/T]TGCCATTTATCACTGTCCTGGCTGC
- Phenotype
-
MIM: 602153
MIM: 601928
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT81
PubMed Links
Gene Details
- Gene
- KRT81
- Gene Name
- keratin 81
There are no transcripts associated with this gene.
- Gene
- KRT86
- Gene Name
- keratin 86
View Full Product Details