Product Details
- SNP ID
-
rs2540054
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:200701247 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGATTTACTGCTTGGATCAAACTC[A/G]GTTCCAATATCGGGGAAATTACTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AOX3P-AOX2P
PubMed Links
Gene Details
- Gene
- AOX3P-AOX2P
- Gene Name
- AOX3P-AOX2P readthrough
There are no transcripts associated with this gene.
View Full Product Details