Product Details

SNP ID
rs8069709
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2416776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCCCGCAACATGAATGTGACCTC[C/T]GGACACCGGGAGAGTAACACTCAAG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LOC284009 PubMed Links

Gene Details

Gene
LOC284009
Gene Name
uncharacterized LOC284009
There are no transcripts associated with this gene.

Gene
METTL16
Gene Name
methyltransferase like 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024086.3 5074 UTR 3 NP_076991.3
XM_005256801.2 5074 UTR 3 XP_005256858.1
XM_017025045.1 5074 UTR 3 XP_016880534.1
XM_017025046.1 5074 UTR 3 XP_016880535.1

View Full Product Details