Product Details

SNP ID
rs6960252
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:77539068 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACATTGGCTAGTTTTTATTTTTT[A/G]TTTAAAAAGTACAGTAGTTTTTATG
Phenotype
MIM: 600079
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPN12 PubMed Links

Gene Details

Gene
PTPN12
Gene Name
protein tyrosine phosphatase, non-receptor type 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131008.1 Intron NP_001124480.1
NM_001131009.1 Intron NP_001124481.1
NM_002835.3 Intron NP_002826.3
XM_005250518.2 Intron XP_005250575.1
XM_006716073.3 Intron XP_006716136.1
XM_011516444.1 Intron XP_011514746.1
XM_017012474.1 Intron XP_016867963.1

View Full Product Details