Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145268.1 | Intron | NP_001138740.1 | |||
NM_001145269.1 | Intron | NP_001138741.1 | |||
XM_006715896.3 | Intron | XP_006715959.1 | |||
XM_011515922.2 | Intron | XP_011514224.1 | |||
XM_011515923.2 | Intron | XP_011514225.1 | |||
XM_011515924.2 | Intron | XP_011514226.1 | |||
XM_011515925.2 | Intron | XP_011514227.1 | |||
XM_011515926.2 | Intron | XP_011514228.1 | |||
XM_017011846.1 | Intron | XP_016867335.1 | |||
XM_017011847.1 | Intron | XP_016867336.1 | |||
XM_017011848.1 | Intron | XP_016867337.1 | |||
XM_017011849.1 | Intron | XP_016867338.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001111038.1 | Intron | NP_001104508.1 | |||
NM_001287150.1 | Intron | NP_001274079.1 | |||
NM_145032.3 | Intron | NP_659469.3 | |||
XM_005250205.3 | Intron | XP_005250262.1 | |||
XM_005250207.3 | Intron | XP_005250264.1 | |||
XM_005250208.3 | Intron | XP_005250265.1 | |||
XM_005250209.2 | Intron | XP_005250266.1 | |||
XM_006715898.2 | Intron | XP_006715961.1 | |||
XM_011515928.2 | Intron | XP_011514230.1 | |||
XM_011515929.2 | Intron | XP_011514231.1 | |||
XM_011515930.2 | Intron | XP_011514232.1 | |||
XM_011515932.2 | Intron | XP_011514234.1 | |||
XM_017011850.1 | Intron | XP_016867339.1 | |||
XM_017011851.1 | Intron | XP_016867340.1 | |||
XM_017011852.1 | Intron | XP_016867341.1 | |||
XM_017011853.1 | Intron | XP_016867342.1 |