Product Details

SNP ID
rs4467294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102741310 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTAGAAACTCTTAGAGAGTTAG[G/T]TTTAGGGATACCCCCACAATTTAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MFSD9 PubMed Links

Gene Details

Gene
MFSD9
Gene Name
major facilitator superfamily domain containing 9
There are no transcripts associated with this gene.

Gene
TMEM182
Gene Name
transmembrane protein 182
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321343.1 Intron NP_001308272.1
NM_001321344.1 Intron NP_001308273.1
NM_001321345.1 Intron NP_001308274.1
NM_001321346.1 Intron NP_001308275.1
NM_144632.4 Intron NP_653233.4
XM_006712287.1 Intron XP_006712350.1
XM_006712288.3 Intron XP_006712351.1
XM_011510632.2 Intron XP_011508934.1
XM_017003375.1 Intron XP_016858864.1
XM_017003376.1 Intron XP_016858865.1

View Full Product Details