Product Details

SNP ID
rs6712162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:36985060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATATACTGCCCATTCTTATACC[A/G]GATACTGTAAAGTTGACAACAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEATR5B PubMed Links

Gene Details

Gene
HEATR5B
Gene Name
HEAT repeat containing 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019024.2 Intron NP_061897.1
XM_006712034.2 Intron XP_006712097.1
XM_006712035.3 Intron XP_006712098.1
XM_011532934.2 Intron XP_011531236.1
XM_011532935.2 Intron XP_011531237.1
XM_017004378.1 Intron XP_016859867.1
XM_017004379.1 Intron XP_016859868.1
XM_017004380.1 Intron XP_016859869.1

View Full Product Details