Product Details

SNP ID
rs414838
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:220692533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATATTATGCTAAGTGAAGTAA[A/G]CAAGACCCGCAAGATCACATATTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf115 PubMed Links

Gene Details

Gene
C1orf115
Gene Name
chromosome 1 open reading frame 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024709.4 Intron NP_078985.3

View Full Product Details