Product Details

SNP ID
rs1277213
Assay Type
Functionally tested
NCBI dbSNP Submissions
71
Location
Chr.1:108816880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTAGAATAATACAAACCTTGCG[A/G]CTGCTGAGGCTGTGTGTCTTTAGCT
Phenotype
MIM: 608339
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKNAD1 PubMed Links

Gene Details

Gene
AKNAD1
Gene Name
AKNA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152763.4 Intron NP_689976.2
Gene
STXBP3
Gene Name
syntaxin binding protein 3
There are no transcripts associated with this gene.

View Full Product Details