Product Details

SNP ID
rs12502513
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7052953 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGGCATGTGTGCCAGAGGAAGC[C/G]CCAGCTGTGGGATCCCATCCGTTCC
Phenotype
MIM: 606173 MIM: 608790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC96 PubMed Links

Gene Details

Gene
CCDC96
Gene Name
coiled-coil domain containing 96
There are no transcripts associated with this gene.

Gene
GRPEL1
Gene Name
GrpE like 1, mitochondrial
There are no transcripts associated with this gene.

Gene
LOC100129931
Gene Name
uncharacterized LOC100129931
There are no transcripts associated with this gene.

Gene
TADA2B
Gene Name
transcriptional adaptor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152293.2 553 Intron NP_689506.2
XM_011513595.1 553 UTR 5 XP_011511897.1

View Full Product Details