Product Details

SNP ID
rs16945091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90955851 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGCAGAGGAGGAGCGCTCAGTA[A/T]ATGCTCGCTGGACGAATAAATGGCT
Phenotype
MIM: 611219
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RCCD1 PubMed Links

Gene Details

Gene
RCCD1
Gene Name
RCC1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017919.1 125 Intron NP_001017919.1
NM_033544.2 125 UTR 5 NP_291022.2
Gene
UNC45A
Gene Name
unc-45 myosin chaperone A
There are no transcripts associated with this gene.

View Full Product Details