Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001143783.1 | Intron | NP_001137255.1 | |||
NM_001143784.1 | Intron | NP_001137256.1 | |||
NM_001143785.1 | Intron | NP_001137257.1 | |||
NM_002005.3 | Intron | NP_001996.1 | |||
XM_005254880.1 | Intron | XP_005254937.1 | |||
XM_005254882.1 | Intron | XP_005254939.1 | |||
XM_017022005.1 | Intron | XP_016877494.1 | |||
XM_017022006.1 | Intron | XP_016877495.1 | |||
XM_017022007.1 | Intron | XP_016877496.1 | |||
XM_017022008.1 | Intron | XP_016877497.1 | |||
XM_017022009.1 | Intron | XP_016877498.1 | |||
XM_017022010.1 | Intron | XP_016877499.1 |