Product Details

SNP ID
rs1565933
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:48862015 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAGTTGATCTAGTCTTACCTTCTT[C/G]AGTGCGCTGTCCACTGTCTCTGGAC
Phenotype
MIM: 612172 MIM: 609038
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX23 PubMed Links

Gene Details

Gene
DDX23
Gene Name
DEAD-box helicase 23
There are no transcripts associated with this gene.

Gene
RND1
Gene Name
Rho family GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014470.3 415 Silent Mutation CTC,CTG L104L NP_055285.1

View Full Product Details