Product Details

SNP ID
rs2040925
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:2912016 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AACAAGGGTGCAGAGAGGGAGTTAA[C/G]ACGGGGCAGCTGCTGGGACAAGGGG
Phenotype
MIM: 607210
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CARD11 PubMed Links

Gene Details

Gene
CARD11
Gene Name
caspase recruitment domain family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324281.1 Intron NP_001311210.1
NM_032415.5 Intron NP_115791.3
XM_011515586.2 Intron XP_011513888.1
XM_011515587.2 Intron XP_011513889.1

View Full Product Details