Product Details

SNP ID
rs34408730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7435581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTAGACAAGGGGCGGCAGCGGA[A/G]GCTGGCTACTGCATTCCGTAGGAGG
Phenotype
MIM: 100710 MIM: 601514 MIM: 613936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf74 PubMed Links

Gene Details

Gene
C17orf74
Gene Name
chromosome 17 open reading frame 74
There are no transcripts associated with this gene.

Gene
CHRNB1
Gene Name
cholinergic receptor nicotinic beta 1 subunit
There are no transcripts associated with this gene.

Gene
FGF11
Gene Name
fibroblast growth factor 11
There are no transcripts associated with this gene.

Gene
LOC107983988
Gene Name
proteoglycan 4-like
There are no transcripts associated with this gene.

Gene
TMEM102
Gene Name
transmembrane protein 102
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320444.1 109 UTR 5 NP_001307373.1
NM_178518.2 109 UTR 5 NP_848613.1

View Full Product Details