Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_013439.2 | 2791 | Intron | NP_038467.2 | ||
NM_178272.1 | 2791 | Intron | NP_840056.1 | ||
NM_178273.1 | 2791 | Intron | NP_840057.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001258008.1 | 2791 | UTR 3 | NP_001244937.1 | ||
NM_017984.4 | 2791 | UTR 3 | NP_060454.3 | ||
XM_005250479.2 | 2791 | UTR 3 | XP_005250536.1 | ||
XM_005250480.2 | 2791 | UTR 3 | XP_005250537.1 | ||
XM_006716035.3 | 2791 | UTR 3 | XP_006716098.1 | ||
XM_006716036.2 | 2791 | UTR 3 | XP_006716099.1 | ||
XM_006716037.3 | 2791 | UTR 3 | XP_006716100.1 | ||
XM_006716038.3 | 2791 | UTR 3 | XP_006716101.1 | ||
XM_006716040.3 | 2791 | UTR 3 | XP_006716103.1 | ||
XM_017012379.1 | 2791 | UTR 3 | XP_016867868.1 | ||
XM_017012380.1 | 2791 | UTR 3 | XP_016867869.1 |