Product Details

SNP ID
rs10790019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114518986 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCACTTTTATTAGTAATGACAGG[C/T]GTATTAATAACAGGGACAAGATTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NXPE1 PubMed Links

Gene Details

Gene
NXPE1
Gene Name
neurexophilin and PC-esterase domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152315.3 5353 Intron NP_689528.2
XM_011542594.2 5353 UTR 3 XP_011540896.1
XM_011542595.2 5353 UTR 3 XP_011540897.1
XM_011542596.2 5353 UTR 3 XP_011540898.1
XM_011542597.2 5353 UTR 3 XP_011540899.1
XM_011542598.2 5353 UTR 3 XP_011540900.1
XM_011542599.2 5353 UTR 3 XP_011540901.1
Gene
NXPE2
Gene Name
neurexophilin and PC-esterase domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182495.5 5353 Intron NP_872301.2
XM_011542604.2 5353 Intron XP_011540906.1
XM_017017206.1 5353 Intron XP_016872695.1
XM_017017207.1 5353 Intron XP_016872696.1
XM_017017208.1 5353 Intron XP_016872697.1
XM_017017209.1 5353 Intron XP_016872698.1
XM_017017210.1 5353 Intron XP_016872699.1
XM_017017211.1 5353 Intron XP_016872700.1
XM_017017212.1 5353 Intron XP_016872701.1

View Full Product Details