Product Details
- SNP ID
-
rs1555079
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
52
- Location
-
Chr.1:11700725 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCGGTTCTCCCCGTGGGGGGTTG[C/T]CTGCTTCACATCATAATCCTTCCGC
- Phenotype
-
MIM: 612682
MIM: 604094
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DRAXIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79991852] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DRAXIN
- Gene Name
- dorsal inhibitory axon guidance protein
- Gene
- MAD2L2
- Gene Name
- MAD2 mitotic arrest deficient-like 2 (yeast)
There are no transcripts associated with this gene.
View Full Product Details