Product Details
- SNP ID
-
rs2233022
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
32
- Location
-
Chr.1:11675820 on Build GRCh38
- Set Membership
-
JSNP
Validated
- Context Sequence [VIC/FAM]
- CTCTTCCCACTTCCCTTGCTGGCTC[A/G]GCAGCACCCCCAGAGCAGATGGCAA
- Phenotype
-
MIM: 605647
MIM: 604094
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2233023] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXO6
- Gene Name
- F-box protein 6
There are no transcripts associated with this gene.
- Gene
- MAD2L2
- Gene Name
- MAD2 mitotic arrest deficient-like 2 (yeast)
View Full Product Details