Product Details

SNP ID
rs2790497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:51850272 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTCTTCCTCTTAAAGTAAGTTG[C/T]CTCTTATGTGGCTGTTAAAATTGCC
Phenotype
MIM: 606981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNG2 PubMed Links

Gene Details

Gene
GNG2
Gene Name
G protein subunit gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243773.1 Intron NP_001230702.1
NM_001243774.1 Intron NP_001230703.1
NM_053064.4 Intron NP_444292.1
XM_006720173.2 Intron XP_006720236.1
XM_011536846.2 Intron XP_011535148.1
XM_017021376.1 Intron XP_016876865.1
XM_017021377.1 Intron XP_016876866.1

View Full Product Details