Product Details
- SNP ID
-
rs2009516
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:94088050 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GATGTAGTCACTCAGCTCTGTTTCA[C/T]GCTCTGCTGTGAACCCTCAAGGATG
- Phenotype
-
MIM: 606181
MIM: 611320
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX24
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4905157,rs75416602] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX24
- Gene Name
- DEAD-box helicase 24
There are no transcripts associated with this gene.
- Gene
- IFI27L1
- Gene Name
- interferon alpha inducible protein 27 like 1
View Full Product Details