Product Details

SNP ID
rs6856731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:181066139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTAACACATACTTTGATTATAG[A/C]CCTGAATTTGACTTGCCACATCACA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00290 PubMed Links

Gene Details

Gene
LINC00290
Gene Name
long intergenic non-protein coding RNA 290
There are no transcripts associated with this gene.

View Full Product Details