Product Details
- SNP ID
-
rs6575304
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:92938329 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CATTAGTGAAGCCATTCAGCAGTTG[C/T]GGCCAGTGGCCAATGTGAGTGCCCA
- Phenotype
-
MIM: 118910
MIM: 601838
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHGA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73345164] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHGA
- Gene Name
- chromogranin A
There are no transcripts associated with this gene.
- Gene
- ITPK1
- Gene Name
- inositol-tetrakisphosphate 1-kinase
View Full Product Details