Product Details

SNP ID
rs1572095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62235396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGCGGGGGAAAGGGCGGTGGGT[C/T]GTAGGCGGGACTGGTGCTGCCAGGC
Phenotype
MIM: 606731
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369209 PubMed Links

Gene Details

Gene
LOC105369209
Gene Name
putative uncharacterized protein FLJ44790
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011529124.1 Intron XP_011527426.1
Gene
OSBPL2
Gene Name
oxysterol binding protein like 2
There are no transcripts associated with this gene.

View Full Product Details