Product Details

SNP ID
rs7552323
Assay Type
Functionally tested
NCBI dbSNP Submissions
56
Location
Chr.1:169134085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTGTGCCAACCTATTTCCCACT[A/C]CTGATCTTTAGTGTTATATCACCAC
Phenotype
MIM: 182330 MIM: 613465
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP1B1 PubMed Links

Gene Details

Gene
ATP1B1
Gene Name
ATPase Na+/K+ transporting subunit beta 1
There are no transcripts associated with this gene.

Gene
NME7
Gene Name
NME/NM23 family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013330.4 Intron NP_037462.1
NM_197972.2 Intron NP_932076.1
XM_005245106.4 Intron XP_005245163.1
XM_017001099.1 Intron XP_016856588.1
XM_017001100.1 Intron XP_016856589.1

View Full Product Details