Product Details

SNP ID
rs1805054
Assay Type
Validated
NCBI dbSNP Submissions
55
Location
Chr.1:19666020 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGCCGGCCATGCTGAACGCGCTGTA[C/T]GGGCGCTGGGTGCTGGCGCGCGGCC
Phenotype
MIM: 601109 MIM: 600613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR6 PubMed Links

Gene Details

Gene
HTR6
Gene Name
5-hydroxytryptamine receptor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000871.2 734 Silent Mutation TAC,TAT Y89Y NP_000862.1
Gene
MINOS1-NBL1
Gene Name
MINOS1-NBL1 readthrough
There are no transcripts associated with this gene.

Gene
NBL1
Gene Name
neuroblastoma 1, DAN family BMP antagonist
There are no transcripts associated with this gene.

View Full Product Details