Product Details

SNP ID
rs4711767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44114250 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAGTCTGCAGTCTGTATCTCT[C/T]ATGGTCAGGGTGCACAGGCCTCTGA
Phenotype
MIM: 611827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL14 PubMed Links

Gene Details

Gene
MRPL14
Gene Name
mitochondrial ribosomal protein L14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318767.1 Intron NP_001305696.1
NM_001318768.1 Intron NP_001305697.1
NM_001318769.1 Intron NP_001305698.1
NM_001318770.1 Intron NP_001305699.1
NM_001318771.1 Intron NP_001305700.1
NM_032111.3 Intron NP_115487.2

View Full Product Details