Product Details
- SNP ID
-
rs11908470
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:35957141 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAATTATTAGAGATAGGAGGAGGG[A/G]CCTGAGAGATCTATTAGAATCATTG
- Phenotype
-
MIM: 610335
MIM: 610416
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CNBD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75090705] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNBD2
- Gene Name
- cyclic nucleotide binding domain containing 2
- Gene
- PHF20
- Gene Name
- PHD finger protein 20
There are no transcripts associated with this gene.
- Gene
- SCAND1
- Gene Name
- SCAN domain containing 1
There are no transcripts associated with this gene.
View Full Product Details