Product Details

SNP ID
rs570710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:801240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTAGCTCCCGCACCCTGAAGAGC[A/C]GGCGTTTCCCAAGCAGAGGGCTCCC
Phenotype
MIM: 612600 MIM: 612895
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRM3 PubMed Links

Gene Details

Gene
MRM3
Gene Name
mitochondrial rRNA methyltransferase 3
There are no transcripts associated with this gene.

Gene
NXN
Gene Name
nucleoredoxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205319.1 Intron NP_001192248.1
NM_022463.4 Intron NP_071908.2
XM_005256756.4 Intron XP_005256813.1
XM_005256758.3 Intron XP_005256815.1
XM_017024949.1 Intron XP_016880438.1

View Full Product Details