Product Details

SNP ID
rs522863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83557253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGCTGGGACTGTGGGGCAGTG[A/T]TGTATATTTTTATTAGTTCAGTGAA
Phenotype
MIM: 607923
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SNAP91 PubMed Links

Gene Details

Gene
SNAP91
Gene Name
synaptosome associated protein 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242792.1 Intron NP_001229721.1
NM_001242793.1 Intron NP_001229722.1
NM_001242794.1 Intron NP_001229723.1
NM_001256717.1 Intron NP_001243646.1
NM_001256718.1 Intron NP_001243647.1
NM_014841.2 Intron NP_055656.1
XM_005248770.4 Intron XP_005248827.1
XM_006715615.1 Intron XP_006715678.1
XM_011536265.1 Intron XP_011534567.1
XM_011536266.1 Intron XP_011534568.1
XM_011536269.1 Intron XP_011534571.1
XM_011536271.1 Intron XP_011534573.1
XM_011536273.2 Intron XP_011534575.1
XM_011536275.2 Intron XP_011534577.1
XM_011536276.2 Intron XP_011534578.1
XM_011536277.1 Intron XP_011534579.1
XM_017011554.1 Intron XP_016867043.1
XM_017011555.1 Intron XP_016867044.1
XM_017011556.1 Intron XP_016867045.1
XM_017011557.1 Intron XP_016867046.1
XM_017011558.1 Intron XP_016867047.1
XM_017011559.1 Intron XP_016867048.1
XM_017011560.1 Intron XP_016867049.1
XM_017011561.1 Intron XP_016867050.1
XM_017011562.1 Intron XP_016867051.1
XM_017011563.1 Intron XP_016867052.1
XM_017011564.1 Intron XP_016867053.1
XM_017011565.1 Intron XP_016867054.1
XM_017011566.1 Intron XP_016867055.1
XM_017011567.1 Intron XP_016867056.1
XM_017011568.1 Intron XP_016867057.1
XM_017011569.1 Intron XP_016867058.1
XM_017011570.1 Intron XP_016867059.1
XM_017011571.1 Intron XP_016867060.1
XM_017011572.1 Intron XP_016867061.1
XM_017011573.1 Intron XP_016867062.1
XM_017011574.1 Intron XP_016867063.1
XM_017011575.1 Intron XP_016867064.1
XM_017011576.1 Intron XP_016867065.1
XM_017011577.1 Intron XP_016867066.1
XM_017011578.1 Intron XP_016867067.1
XM_017011579.1 Intron XP_016867068.1
XM_017011580.1 Intron XP_016867069.1
XM_017011581.1 Intron XP_016867070.1
XM_017011582.1 Intron XP_016867071.1
XM_017011583.1 Intron XP_016867072.1
XM_017011584.1 Intron XP_016867073.1
XM_017011585.1 Intron XP_016867074.1
XM_017011586.1 Intron XP_016867075.1
XM_017011587.1 Intron XP_016867076.1
XM_017011588.1 Intron XP_016867077.1
XM_017011589.1 Intron XP_016867078.1
XM_017011590.1 Intron XP_016867079.1

View Full Product Details