Product Details
- SNP ID
-
rs7767309
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:106366092 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTCTTCAGAAAAAGAACAATCAT[C/T]TAAAATGTTATTTGAAAGCATTCTG
- Phenotype
-
MIM: 601797
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AIM1
PubMed Links
Gene Details
- Gene
- AIM1
- Gene Name
- absent in melanoma 1
View Full Product Details