Product Details

SNP ID
rs10193011
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:6919801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTTTGTGTTTTTAAAGAGCTCC[C/T]GATTCTAATGTCAGCAACTCTGGAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RNF144A PubMed Links

Gene Details

Gene
RNF144A
Gene Name
ring finger protein 144A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014746.4 Intron NP_055561.2
XM_005246200.3 Intron XP_005246257.1
XM_005246202.4 Intron XP_005246259.1
XM_017005396.1 Intron XP_016860885.1
XM_017005397.1 Intron XP_016860886.1
XM_017005398.1 Intron XP_016860887.1
XM_017005399.1 Intron XP_016860888.1
XM_017005400.1 Intron XP_016860889.1
XM_017005401.1 Intron XP_016860890.1
XM_017005402.1 Intron XP_016860891.1
XM_017005403.1 Intron XP_016860892.1
XM_017005404.1 Intron XP_016860893.1
Gene
RNF144A-AS1
Gene Name
RNF144A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details