Product Details
- SNP ID
-
rs6731402
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:113711558 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAAATTGCAAGTCTATGTGAAAAAT[C/G]AACCTCAATTTATCTAAATGGGATT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4782
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11298773] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4782
- Gene Name
- microRNA 4782
There are no transcripts associated with this gene.
- Gene
- SLC35F5
- Gene Name
- solute carrier family 35 member F5
View Full Product Details