Product Details

SNP ID
rs283891
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:79122246 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCAGAAATCAGGCTGTTTACAGAT[C/G]CTCTAATGTCCTGTGTAGCAAGGTG
Phenotype
MIM: 167770
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
REG1A PubMed Links

Gene Details

Gene
REG1A
Gene Name
regenerating family member 1 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002909.4 Intron NP_002900.2

View Full Product Details