Product Details

SNP ID
rs13225906
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:114928049 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGGATTTGGGAACATTTTGAATTT[A/G]GAAAATGAATCTTAGAACTTATACT
Phenotype
MIM: 614511
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MDFIC PubMed Links

Gene Details

Gene
MDFIC
Gene Name
MyoD family inhibitor domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166345.1 Intron NP_001159817.1
NM_001166346.1 Intron NP_001159818.3
NM_199072.4 Intron NP_951038.1

View Full Product Details