Product Details
- SNP ID
-
rs10196645
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:33726446 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCCGCTCACTGGAAATTATTGAC[C/T]TCCAGAGATACATACATTTTTTTTA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01317
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114331606] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01317
- Gene Name
- long intergenic non-protein coding RNA 1317
There are no transcripts associated with this gene.
- Gene
- MYADML
- Gene Name
- myeloid associated differentiation marker like (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details