Product Details

SNP ID
rs7649480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119301578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGTGGAAGCAGGTGTGAGGGGA[A/C]ACACATTCCGAATTCCAAAAGTTGA
Phenotype
MIM: 610911
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP31 PubMed Links

Gene Details

Gene
ARHGAP31
Gene Name
Rho GTPase activating protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020754.3 Intron NP_065805.2
XM_006713714.3 Intron XP_006713777.1
XM_017006955.1 Intron XP_016862444.1

View Full Product Details