Product Details

SNP ID
rs2167079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47248704 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAACGAAGCGCAGACTCCGGGCC[C/T]GGGTGGGCGGCATCACCACCAGGTT
Phenotype
MIM: 171650 MIM: 600811 MIM: 602423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACP2 PubMed Links

Gene Details

Gene
ACP2
Gene Name
acid phosphatase 2, lysosomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302489.1 81 Intron NP_001289418.1
NM_001302490.1 81 Missense Mutation CAG,CGG Q29R NP_001289419.1
NM_001302491.1 81 Missense Mutation CAG,CGG Q29R NP_001289420.1
NM_001302492.1 81 UTR 5 NP_001289421.1
NM_001610.3 81 Missense Mutation CAG,CGG Q29R NP_001601.1
Gene
DDB2
Gene Name
damage specific DNA binding protein 2
There are no transcripts associated with this gene.

Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130101.2 81 Intron NP_001123573.1
NM_001130102.2 81 Intron NP_001123574.1
NM_001251934.1 81 UTR 5 NP_001238863.1
NM_001251935.1 81 UTR 5 NP_001238864.1
NM_005693.3 81 Intron NP_005684.2
XM_005252705.1 81 Intron XP_005252762.1
XM_005252706.1 81 Intron XP_005252763.1
XM_005252707.4 81 Intron XP_005252764.1
XM_005252709.1 81 Intron XP_005252766.1
XM_005252710.1 81 Intron XP_005252767.1
XM_005252713.3 81 Intron XP_005252770.1
XM_005252715.2 81 Intron XP_005252772.1
XM_005252716.3 81 Intron XP_005252773.1
XM_005252718.3 81 Intron XP_005252775.1
XM_006718112.1 81 Intron XP_006718175.1
XM_006718113.1 81 Intron XP_006718176.1
XM_006718115.1 81 Intron XP_006718178.1
XM_006718116.1 81 Intron XP_006718179.1
XM_011519805.2 81 Intron XP_011518107.1
XM_011519806.1 81 Intron XP_011518108.1
XM_011519807.1 81 Intron XP_011518109.1
XM_011519808.2 81 Intron XP_011518110.1
XM_017017056.1 81 Intron XP_016872545.1
XM_017017057.1 81 Intron XP_016872546.1
XM_017017058.1 81 Intron XP_016872547.1

View Full Product Details