Product Details

SNP ID
rs3792345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:146071847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAACAAGCTTTCCCCGCCCTTC[C/T]TCACTTCTATTTGTAACACTCAAAA
Phenotype
MIM: 601865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLOD2 PubMed Links

Gene Details

Gene
PLOD2
Gene Name
procollagen-lysine,2-oxoglutarate 5-dioxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000935.2 Intron NP_000926.2
NM_182943.2 Intron NP_891988.1
XM_005247535.3 Intron XP_005247592.1
XM_017006625.1 Intron XP_016862114.1

View Full Product Details