Product Details

SNP ID
rs1294308
Assay Type
Functionally tested
NCBI dbSNP Submissions
57
Location
Chr.1:233331872 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACAACTTTGTCCCTTTTCTAACA[A/G]GTAGCAACAGGTTGAAAGTTGTGCC
Phenotype
MIM: 614793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1804 PubMed Links

Gene Details

Gene
KIAA1804
Gene Name
mixed lineage kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032435.2 Intron NP_115811.2
XM_011544305.2 Intron XP_011542607.1

View Full Product Details