Product Details
- SNP ID
-
rs7601782
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:11156366 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCAAAGTATTCATCATGCCAGAGC[C/G]CCAAACTTTTGGGGTATCATTTTTG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C2orf50
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77173613] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2orf50
- Gene Name
- chromosome 2 open reading frame 50
There are no transcripts associated with this gene.
- Gene
- PQLC3
- Gene Name
- PQ loop repeat containing 3
View Full Product Details