Product Details
- SNP ID
-
rs34677348
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39352645 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGTTGGCCTGCTTTGTTCAAAGC[C/G]TGGCCTTGTAAGCATCATGTTTGAG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SAMD4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75372153] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SAMD4B
- Gene Name
- sterile alpha motif domain containing 4B
View Full Product Details