Product Details

SNP ID
rs939649
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:44590720 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAAAAGAGAAGTTCCAGAGAAGAT[G/T]AGAAATTGAAAAGAAATTCAGAAAA
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZKSCAN7 PubMed Links

Gene Details

Gene
ZKSCAN7
Gene Name
zinc finger with KRAB and SCAN domains 7
There are no transcripts associated with this gene.

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 196 Intron NP_775929.2
XM_005265090.4 196 UTR 5 XP_005265147.1
XM_006713117.3 196 Intron XP_006713180.1
XM_011533622.2 196 Intron XP_011531924.1

View Full Product Details