Product Details
- SNP ID
-
rs11160255
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:95536732 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AAATGGTAGCTGGCTTCGTATTAGC[A/C]CTTGATGGTTTTCAAATGTATTTTT
- Phenotype
-
MIM: 609588
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GLRX5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79171925] are located under a probe and SNP(s) [rs112951493] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLRX5
- Gene Name
- glutaredoxin 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016417.2 |
|
Intron |
|
|
NP_057501.2 |
- Gene
- SCARNA13
- Gene Name
- small Cajal body-specific RNA 13
There are no transcripts associated with this gene.
- Gene
- SNHG10
- Gene Name
- small nucleolar RNA host gene 10
There are no transcripts associated with this gene.
View Full Product Details