Product Details

SNP ID
rs4905335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95538009 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAACTTATGCTCATGGAAGCAA[C/T]GTAGAAAACACTGAAAAGTTTAAAG
Phenotype
MIM: 609588
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLRX5 PubMed Links

Gene Details

Gene
GLRX5
Gene Name
glutaredoxin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016417.2 Intron NP_057501.2
Gene
SCARNA13
Gene Name
small Cajal body-specific RNA 13
There are no transcripts associated with this gene.

Gene
SNHG10
Gene Name
small nucleolar RNA host gene 10
There are no transcripts associated with this gene.

View Full Product Details