Product Details

SNP ID
rs12419480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95978168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAAAGTGAAATCTAATGCAGA[C/G]AGGTGTGCACCAATATTTCAGTATG
Phenotype
MIM: 607537
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAML2 PubMed Links

Gene Details

Gene
MAML2
Gene Name
mastermind like transcriptional coactivator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032427.3 7371 UTR 3 NP_115803.1
XM_011543023.2 7371 UTR 3 XP_011541325.1
XM_011543024.2 7371 UTR 3 XP_011541326.1
XM_011543025.1 7371 Intron XP_011541327.1

View Full Product Details