Product Details
- SNP ID
-
rs1719581
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:189963769 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACAATTTGTTAATAAGCTCAAGA[C/G]AGATTAGTGACTATCAAATATCATT
- Phenotype
-
MIM: 610341
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
P3H2
PubMed Links
Gene Details
- Gene
- P3H2
- Gene Name
- prolyl 3-hydroxylase 2
View Full Product Details