Product Details

SNP ID
rs8079818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63747307 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTCACCATACCATTCATGATGG[C/G]ATTATATCATAAATGTATAACTGCC
Phenotype
MIM: 608626
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC47 PubMed Links

Gene Details

Gene
CCDC47
Gene Name
coiled-coil domain containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020198.2 Intron NP_064583.2
XM_005257527.2 Intron XP_005257584.1
Gene
STRADA
Gene Name
STE20-related kinase adaptor alpha
There are no transcripts associated with this gene.

View Full Product Details