Product Details

SNP ID
rs2279254
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:70432214 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTAACAAACTCAGGCTGTGTCTGC[C/G]GAAGCAGACTGACGCAGGGATGTCT
Phenotype
MIM: 602224 MIM: 601265
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EIF4EBP2 PubMed Links

Gene Details

Gene
EIF4EBP2
Gene Name
eukaryotic translation initiation factor 4E binding protein 2
There are no transcripts associated with this gene.

Gene
NODAL
Gene Name
nodal growth differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018055.4 1808 UTR 3 NP_060525.3

View Full Product Details