Product Details

SNP ID
rs12670233
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:32505978 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAAAGATTCTAAGTCCGTAAAGTGT[A/G]CCCTAAACATAATCTCTGCTTGTTA
Phenotype
MIM: 612927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AVL9 PubMed Links

Gene Details

Gene
AVL9
Gene Name
AVL9 cell migration associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015060.2 Intron NP_055875.1
XM_005249668.2 Intron XP_005249725.1
XM_005249669.2 Intron XP_005249726.1
XM_005249670.2 Intron XP_005249727.1
XM_005249671.4 Intron XP_005249728.1
XM_011515227.2 Intron XP_011513529.1
XM_017011891.1 Intron XP_016867380.1
XM_017011892.1 Intron XP_016867381.1

View Full Product Details